missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Canine IgE Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen SA510298
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Stable for a minimum of 1 year at -20°C.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Canine IgE | |
Monoclonal | |
3.84 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Canine IgE | |
1 mg | |
Secondary | |
4° C | |
IgG1 |
ELISA, Western Blot | |
2A2 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Canine | |
Liquid |