missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Hamster IgG1, Clone: G94-56, NA/LE, Secondary Antibody, BD

Brand:  BD Biosciences 554005

295.87 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15897528

  • 296.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Based on ELISA, the G94-56 antibody reacts specifically with Armenian and Syrian hamster IgG1 monoclonal antibodies, but has low reactivity with some other hamster IgG groups. (Please refer to the Reactivity of Mouse Anti-Hamster Ig mAbs chart on our website for more information: http://www.bdbiosciences.com/pharmingen/hamster_chart_11x17.pdf.) The G94-56 mAb does not react with hamster IgM.

Host Species: Mouse
Clone: G94-56
Isotype: IgG2b, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian hamster IgG mAb

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse anti-Hamster IgG1
G94-56
Mouse
0.5 mg
Immunology
Hamster
Low Endotoxin/Azide-free (LE/AF)
1mg/mL
Affinity Purified
RUO
Secondary
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mouse anti-Hamster IgG1, Clone: G94-56, NA/LE, Secondary Antibody, BD > 0.5mg; Unlabeled, NA/LE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.