Learn More
Mouse anti-Human IgA1/IgA2, Clone: G18-1, Secondary Antibody, BD
Brand: BD Biosciences 555886
192.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The G18-1 antibody reacts specifically with human IgA1 and IgA2. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G18-1
Isotype: IgG1
ELISA

Specifications
Mouse anti-Human IgA1/IgA2 | |
G18-1 | |
Mouse | |
0.5 mg | |
Immunology | |
Human |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 κ |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.