missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgA1/IgA2, Clone: G20-359, Biotin, Secondary Antibody, BD

Brand:  BD Biosciences 555884

230.17 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15838508

  • 231.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The G20-359 antibody reacts specifically with human IgA1 and IgA2. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: G20-359
Isotype: IgG1, Îş
Species Reactivity: Human

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse anti-Human IgA1/IgA2
G20-359
Biotin
0.5 mg
Immunology
Human
Monoclonal
0.5mg/mL
Mouse
RUO
Secondary
IgG1 Îş
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mouse anti-Human IgA1/IgA2, Clone: G20-359, Biotin, Secondary Antibody, BD > 0.5mg; Biotin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.