missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA135469
This item is not returnable.
View return policy
Description
This antibody does not cross-react with HSA or other albumins.
Human IgG (H+L) Monoclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Human IgG (H+L) | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5 | |
Purified human IgG. | |
1 mL | |
Secondary | |
-20° C, store in dark | |
IgG1 |
ELISA, Immunohistochemistry | |
EFE-565 | |
FITC | |
Mouse | |
purified | |
RUO | |
Human | |
Liquid |