missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG4, Clone: G17-4, Secondary Antibody, BD
Brand: BD Biosciences 555881
This item is not returnable.
View return policy
Description
The G17-4 antibody reacts specifically with human IgG4. It does not react with other isotypes.
Host Species: Mouse
Clone: G17-4
Isotype: IgG1, κ
Species Reactivity: Human
ELISA
Specifications
Mouse anti-Human IgG4 | |
G17-4 | |
Mouse | |
0.5 mg | |
Immunology | |
Human |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 κ |