missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse brain tissue lysate, Non-denatured; Abnova
Brand: Abnova L035W2.200ug
This item is not returnable.
View return policy
Description
- Tissue: Brain
- Host: Mouse
- Lysis buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Specifications
Immunoprecipitation, Western Blot | |
Whole tissue lysate (non-denatured) | |
Tissue lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was adjusted to 5 mg/ml. | |
200 μg | |
Mouse |
Brain | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In modified RIPA Lysis Buffer. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse brain tissue lysate, Non-denatured; Abnova > 200μg
Spot an opportunity for improvement?Share a Content Correction