missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB22972-SP
This item is not returnable.
View return policy
Description
IL-36 alpha/IL-1F6 Monoclonal antibody specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for ELISA
Specifications
IL-36 alpha/IL-1F6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
E. coli-derived recombiMouset mouse IL-36 alpha/IL-1F6, Met1-His160, Accession # Q9JLA2 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2a |
ELISA | |
275302 | |
ELISA | |
Q9JLA2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27179 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™ > 25 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction