missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-5 Quantikine ELISA Kit
Mouse IL-5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
923.30€ - 2629.52€
Specifications
Assay Range | 15.60 - 1000pg/mL |
---|---|
Assay Sensitivity | 7pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Immunoassay Kit Format | 96-well Plate |
Product Code | Brand | Quantity | Immunoassay Kit Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Immunoassay Kit Format | Price | Quantity & Availability | ||||
18791794
|
R&D Systems™
SM5000 |
1 Pack | 96-well Plate |
2629.52€
1 set |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
18732424
|
R&D Systems™
M5000 |
1 Kit | 96-well Plate |
923.30€
1 set |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
Description
The Mouse IL-5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-5 in cell culture supernates (50 ul), serum (50 ul).Specifications
15.60 - 1000pg/mL | |
HRP | |
96-well Plate | |
Natural and Recombinant Mouse IL-5 | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL) | |
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5 T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I |
7pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant, Serum | |
Mouse | |
16191 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |