missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse NCAM-1/CD56 (NP_001106675) VersaClone cDNA
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Brand: R&D Systems™ RDC2716
This item is not returnable.
View return policy
Description
- Also known as: CD56; Ncam; ENCAM; NCAM-1
- NCAM1 is a cell adhesion protein and a member of the immunoglobulin superfamily.
- It is involved in cell-to-cell interactions as well as cell-matrix interactions during development and differentiation.
- NCAM1 has been shown to be involved in development of the nervous system, and for cells involved in the expansion of T cells and dendritic cells which play an important role in immune surveillance.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
mNcam1 | |
10 μg at 0.2 μg/μL | |
3361 bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_001106675 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
4684 | |
Mouse |