Learn More
R&D Systems™ Mouse Thrombomodulin/BDCA-3 (NP_033404.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3400
This item is not returnable.
View return policy
Specifications
Thrombomodulin/BDCA-3 | |
7056 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_033404.1 | |
BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM | |
Mouse |