Learn More
R&D Systems™ Mouse XL LIF, Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems™ LMXL8878
Product Code. 18491900
  • 227.00€ / 1 set
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
1 Kit
3976
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Mouse
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI