missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Swine, Purified, 2mL

Whole Swine Serum with buffer salts

Brand:  MP Biomedicals™ 0855993

Product Code. 11404325

  • 42.40€ / 2mL
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Swine
Swine Serum
60.3 mg/mL
Serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
Lyophilized
2 mL
Dialyzed into Sodium Phosphate 0.02M, Sodium Chloride 0.14M, pH 7.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers