missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTH1 Rabbit anti-Human, Clone: 0P6T10, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00€ - 494.00€
Specifications
Antigen | MTH1 |
---|---|
Clone | 0P6T10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18333293
|
Bio-Techne
NBP3-16297-20UL |
20 μg |
196.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392587
|
Bio-Techne
NBP3-16297-100UL |
100 μg |
494.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
MTH1 Monoclonal antibody specifically detects MTH1 in Human samples. It is validated for Western BlotSpecifications
MTH1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4521 | |
IgG | |
Affinity purified |
0P6T10 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human MTH1 (P36639). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title