Learn More
Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES
Perform susceptibility testing with a full range of supplies, including this broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Brand: Thermo Scientific™ YT3462-05
380.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform high-quality susceptibility testing with a complete range of supplies, including Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES | |
Microbial Susceptibility | |
5 mL/Bottle |
Microbiology Media | |
100 | |
100 x 5 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
255970 | Certificate of Analysis | 05/05/2025 | YT3462-05 |
252849 | Certificate of Analysis | 13/04/2025 | YT3462-05 |
244200 | Certificate of Analysis | 21/03/2025 | YT3462-05 |
227753 | Certificate of Analysis | 14/02/2025 | YT3462-05 |
223157 | Certificate of Analysis | 30/01/2025 | YT3462-05 |