Learn More
Multi-Element Organometallic Oil Standards 23 Element, 900μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
462.00€ - 831.00€
Specifications
Chemical Name or Material | 23 Element Oil Standard |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Analyte or Component Names | Al,Ba,B |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11460359
|
SPEX Certiprep
S23-900Z |
50 g | Triple-leached LDPE bottle |
462.00€
50g |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
15246386
|
SPEX Certiprep
S23-900Y |
100 g | Triple-leached LDPE bottle |
831.00€
100g |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 900μg/g |
Al | 900μg/g |
B | 900μg/g |
Ba | 900μg/g |
Ca | 900μg/g |
Cd | 900μg/g |
Cr | 900μg/g |
Cu | 900μg/g |
Fe | 900μg/g |
K | 900μg/g |
Mg | 900μg/g |
Mn | 900μg/g |
Mo | 900μg/g |
Na | 900μg/g |
Ni | 900μg/g |
P | 900μg/g |
Pb | 900μg/g |
Sb | 900μg/g |
Si | 900μg/g |
Sn | 900μg/g |
Ti | 900μg/g |
V | 900μg/g |
Zn | 900μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
23 Element Oil Standard | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Al,Ba,B |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Base Oil 75 |
Safety and Handling
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.