Learn More
Multi-Element QC-21 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
435.17€ - 1005.00€
Specifications
Chemical Name or Material | Multi-Element QC-21 Check Standard Solution |
---|---|
Concentration | Matrix: 5% HNO3/tr. F-/tr. tartaric acid |
MDL Number | MFCD00803610 |
UN Number | UN3264 |
Grade | Specpure™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11364038
|
Thermo Scientific Alfa Aesar
041328.AE |
100 mL |
435.17€
100mL |
In Stock
Log in to see stock. |
|||||
11490381
|
Thermo Scientific Alfa Aesar
041328.AP |
500 mL |
1005.00€
500mL |
In Stock
Log in to see stock. |
|||||
Description
Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Multi-Element QC-21 Check Standard Solution | |
MFCD00803610 | |
Specpure™ | |
Liquid |
Matrix: 5% HNO3/tr. F-/tr. tartaric acid | |
UN3264 | |
Miscible with water |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only