Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Accessory, Manifold
Accurate and precise dispensing
Brand: Agilent BioTek 7180560S
This item is not returnable.
View return policy
Description
- Syringe Dispenser Manifold
- 96-/384-well Plates - 2x8-tube
- Autoclavable
Specifications
Half-area and PCR 96-well formats,Steel tips | |
Steel | |
Autoclavable |
CE and TUV marked. RoHS compliant. | |
2 x 8 Manifold | |
Multiflo FX, 96- and 384-well Dispensing |