Learn More
Hanna Instruments™ Multiparameter Photometer
Brand:  Hanna Instruments™ HI8321402
 View more versions of this product
Product Code. 11568039
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Specifications
Multiparameter Photometer
USB
115/230 VAC
235 mm
0.9 kg
LCD
Wastewater Analysis
110 mm
Tungsten lamps with different narrow band interference filters
200 mm