Learn More
Cytiva Multiphor II Buffer Strip Positioner, complete
Multiphor II Buffer Strip Positioner is a frame with slots that sits on top of ExcelGel precast gel after the gel is placed on Multiphor II cooling plate.
Brand:  Cytiva 80-6442-90
Product Code. 10279104
  • 559.00€ / Each
Estimated Shipment: 22-01-2025
Description
The slots in the positioner facilitate placement of the buffer strips for electrophoresis and hold the strips securely in place during the run.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Only for use with Multiphor II cooling plate
Multiphor II Buffer Strip Positioner