Learn More
Thermo Scientific Chemicals Myelin Oligodendrocyte Glycoprotein Peptide (35-55), Rat, Mouse
For cellular and molecular biology applications
181.00€ - 284.42€
Specifications
Species | Mouse, Rat |
---|---|
Purification Method | HPLC |
Common Name | MOG Peptide (35-55) |
Content And Storage | -30°C to -10°C |
Format | Powder |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mouse, Rat | |
MOG Peptide (35-55) | |
Powder | |
Myelin Oligodendrocyte Glycoprotein Peptide (35-55), rat, mouse | |
Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys |
HPLC | |
-30°C to -10°C | |
2582.00 | |
RUO - research use only |