Learn More
Novus Biologicals™ Myelin Protein Zero Overexpression Lysate
Brand:  Novus Biologicals™ NBL1-13213
Product Code. 18202151
  • 700.00€ / 0.10mg
Description
The Myelin Protein Zero Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC2245 and based on accession number NM_53. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens myelin protein zero (MPZ), mRNA.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Myelin Protein Zero Overexpression Lysate > 0.1mg