Learn More
Thermo Scientific Chemicals N-Acetyl-L-proline, 99% Product Code.: 11303456
Thermo Scientific Chemicals N-Acetyl-L-proline, 99%
Product Code. 11303456
5 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 11303456
Brand: Thermo Scientific Chemicals L14300.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68-95-1
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00020837
InChI Key GNMSLDIYJOSUSW-UHFFFAOYNA-N
Synonym n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acidShow MoreShow Less
PubChem CID 66141
ChEBI CHEBI:21560
IUPAC Name (2S)-1-acetylpyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CCCC1C(O)=O
Specifications
Melting Point 117°C to 118°C
Beilstein 83200
Quantity 5 g
Formula Weight 157.18
Optical Rotation −116° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-proline
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249993 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberL14300.06,L14300.03
Lot Number10217864 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog NumberL14300.06,L14300.03
Lot NumberW27E051 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14300.06,L14300.03
Lot NumberZ16F025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14300.06,L14300.03
Lot Number10176667 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14300.06,L14300.03
Product Title
Thermo Scientific Chemicals N-Acetyl-L-proline, 99% >