missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylmaleamic acid, 98+%, Thermo Scientific™

Product Code. 11415387
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15329-69-8
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00020482
InChI Key BHWGQIYJCMMSNM-SREVYHEPSA-N
Synonym n-benzylmaleamic acid, z-4-benzylamino-4-oxobut-2-enoic acid, 2z-3-benzylcarbamoyl prop-2-enoic acid, 2e-4-benzylamino-4-oxobut-2-enoic acid, n-benzyl maleamic acid, maleic acid monobenzylamide, z-3-benzylcarbamoyl-acrylic acid, 2z-3-n-benzylcarbamoyl prop-2-enoic acid, 2z-4-benzylamino-4-oxobut-2-enoic acid
PubChem CID 1549764
IUPAC Name (Z)-4-(benzylamino)-4-oxobut-2-enoic acid
SMILES C1=CC=C(C=C1)CNC(=O)C=CC(=O)O

Specifications

Melting Point 136°C to 138°C
Quantity 5 g
Beilstein 1212733
Formula Weight 205.21
Percent Purity ≥98%
Chemical Name or Material N-Benzylmaleamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'