missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-3,4-dichloro-D-phenylalanine, 95%, Thermo Scientific™ Product Code.: 15478587

N-Boc-3,4-dichloro-D-phenylalanine, 95%, Thermo Scientific™

Product Code. 15478587
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15478587

Brand: Thermo Scientific Alfa Aesar H52070.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114873-13-1
Molecular Formula C14H16Cl2NO4
Molecular Weight (g/mol) 333.19
MDL Number MFCD00151887
InChI Key UGZIQCCPEDCGGN-LLVKDONJSA-M
Synonym boc-3,4-dichloro-d-phenylalanine, boc-d-phe 3,4-cl2-oh, r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-d-3,4-dichlorophe, boc-d-3,4-dichlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-d-phe 3,4-dicl-oh, boc-3,4-dichloro-d-phe-oh, n-boc-d-3,4-dichlorophenylalanine, d-phenylalanine, 3,4-dichloro-n-1,1-dimethylethoxy carbonyl
PubChem CID 7015811
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O

Specifications

Quantity 1 g
Beilstein 8428163
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-dichlorophenyl)propanoate
Formula Weight 334.2
Percent Purity 95%
Chemical Name or Material N-Boc-3,4-dichloro-D-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ20E032 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberK19W082 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberZ04B047 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-3,4-dichloro-D-phenylalanine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.