missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-cyclohexyl-L-alanine, 98%, Thermo Scientific Chemicals

Product Code. 15471488
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37736-82-6
Molecular Formula C14H25NO4
Molecular Weight (g/mol) 271.357
MDL Number MFCD00211335
InChI Key MSZQAQJBXGTSHP-NSHDSACASA-N
Synonym boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine
PubChem CID 10588189
IUPAC Name (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O

Specifications

Quantity 250 mg
Beilstein 3060603
Formula Weight 271.36
Percent Purity 98%
Chemical Name or Material N-Boc-3-cyclohexyl-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'