missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10578030
Click to view available options
Quantity:
500 mL
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Butyl Acetate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Weight (g/mol) 116.16
MDL Number 9445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -90°C
Color Colorless
pH 6.2
Boiling Point 126°C
Packaging Amber glass bottle
Molecular Formula C6H12O2
Quantity 500 mL
Vapor Density 4
Formula Weight 116.16g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.