missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N-Methyl-L-arginine is used as a selective inhibitor of L-arginine uptake. It comes under the category of ion pump transporters.
Solubility
Soluble to 100 mM in water
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. For obtaining a higher solubility , please warm the tube at 37°C and shake it in the ultrasonic bath for a while. Stock solution can be stored below -20°C for several months.
N-Methyl-L-arginine is used as a selective inhibitor of L-arginine uptake. It comes under the category of ion pump transporters.
Solubility
Soluble to 100 mM in water
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. For obtaining a higher solubility , please warm the tube at 37°C and shake it in the ultrasonic bath for a while. Stock solution can be stored below -20°C for several months.

Specifications
Specifications
Color | White |
Solubility Information | Soluble to 100 mM in water |
Quantity | 10 mg |
Formula Weight | 188.23 |
Physical Form | Powder |
Chemical Name or Material | N-Methyl-L-arginine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Methyl-L-arginine, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction