missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals

CAS: 611-74-5 | C9H11NO | 149.19 g/mol

45.55€ - 522.00€

Chemical Identifiers

CAS 611-74-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00008320
InChI Key IMNDHOCGZLYMRO-UHFFFAOYSA-N
Synonym dimethylbenzamide, benzamide, n,n-dimethyl, nn-dimethylbenzamide, dimethylbenzmide, n,n-dmethylbenzamde, dmbz, n,n-dimethyl benzamide, n,n-dimethyl-benzamide, acmc-209mps, benzamide,n,n-dimethyl
PubChem CID 11916
IUPAC Name N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493038
SDS View Documents
Thermo Scientific Alfa Aesar
A18766.14
25 g
45.55€
25g
Please to purchase this item. Need a web account? Register with us today!
11403048
SDS View Documents
Thermo Scientific Alfa Aesar
A18766.22
100 g
131.00€
100g
Please to purchase this item. Need a web account? Register with us today!
11413048
SDS View Documents
Thermo Scientific Alfa Aesar
A18766.36
500 g
522.00€
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reagent for deoxygenation of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for deoxygenation of secondary alcohols.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.