Learn More
n-Octylbenzene, 99%, Thermo Scientific Chemicals
Description
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -36°C |
Density | 0.854 |
Boiling Point | 264°C to 265°C |
Flash Point | 107°C (224°F) |
Odor | Mild |
Refractive Index | 1.484 |
Quantity | 100 g |
Beilstein | 1906253 |
Solubility Information | Immiscible with water. |
Formula Weight | 190.33 |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.