Learn More
Macherey-Nagel™ N9 Polypropylene Red Screw Caps
Brand:  Macherey-Nagel™ 702037
Product Code. 14843532
  • 26.30€ / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Red
Silicone white/PTFE red
N9
Polypropylene, PTFE, Silicone
1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ N9 Polypropylene Red Screw Caps > Septum: Silicone white/PTFE red