Learn More
NAG Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-18441AF700
Product Code. 18596249
  • 663.00€ / 0.10mL
Explore more special offers
Description
Description
NAG Polyclonal antibody specifically detects NAG in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NAG
Polyclonal
Western Blot
DKFZp586G1219, EC 1.1.1.94, EC 1.2.1.59, FLJ40407, NAG/BC035112 fusion, NAG/FAM49A fusion, NAGneuroblastoma-amplified protein, neuroblastoma amplified sequence, Neuroblastoma-amplified gene protein, neuroblastoma-amplified sequence
Recognizes region between residues Between Between 350 and 400..
0.1 mL
Cell Biology
51594
Store at 4°C in the dark.
IgG
Western Blot
Alexa Fluor 700
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
NAG Antibody, Alexa Fluor™ 700, Novus Biologicals™ > 0.1 mL; Alexa Fluor 700