Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners, 200 gallon
Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Brand:  Thermo Scientific™ 343050-0200
1314.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10243243
  • 1470.00€ / Pack of 10
Estimated Shipment: 24-04-2025
Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks. Coex Polyethylene film is free of animal-derived components (ADCF).
  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Product Title
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners, 200 gallon > Gamma Irradiated: 200 gal. (757L)