missing translation for 'onlineSavingsMsg'
Learn More
Naphthalene, 99+%, Thermo Scientific Chemicals Product Code.: 11414594

Naphthalene, 99+%, Thermo Scientific Chemicals

Product Code. 11414594
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11414594

Brand: Thermo Scientific Alfa Aesar A13188.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-20-3 | C10H8 | 128.17 g/mol

Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

Solubility
Soluble in benzene, toluene, chloroform, carbon disulfide, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 80°C to 82°C
Density 0.963
Boiling Point 218°C
Flash Point 78°C (172°F)
Odor Aromatic
Refractive Index 1.5821
Quantity 1000 g
UN Number UN1334
Beilstein 1421310
Merck Index 14,6370
Solubility Information Soluble in benzene,toluene,chloroform,carbon disulfide,ether. Insoluble in water.
Formula Weight 128.17
Percent Purity ≥99%
Chemical Name or Material Naphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460958 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberA13188.0B,A13188.30,A13188.0I
Lot NumberA0460959 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberA13188.0B,A13188.30,A13188.0I
Lot NumberA0446834 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA13188.0B,A13188.30,A13188.0I
Lot NumberA0429345 Certificate TypeCertificate of Analysis Date19/10/2022 Catalog NumberA13188.0B,A13188.30,A13188.0I
Lot NumberA0429435 Certificate TypeCertificate of Analysis Date19/10/2022 Catalog NumberA13188.0B,A13188.30,A13188.0I

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Naphthalene, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.