Learn More
BRAND™ Narrow Neck Erlenmeyer Flask
Narrow Neck Erlenmeyer Flask features a beaded rim and graduation marks.
Brand: BRAND™ 92773
103.89 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate Glass 3.3
- DIN ISO 1773
- With beaded rim and graduation
Specifications
5000 mL | |
Narrow | |
Beaded |
Borosilicate Glass (3.3) | |
Yes |