missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Natural Rubber Stoppers
Made from elastic natural rubber, these stoppers are flexible and provide a complete seal and good adhesion. Saint-Gobain Performance Plastics™ Natural Rubber Stoppers withstand chemical agents in normal laboratory use. Ideal for use with pipets, vials, bottles, flasks or balloons.
Brand: Saint-Gobain 31D203031
This item is not returnable.
View return policy
Description
- Temperature resistance from -35 to +70°C (120°C in humid conditions)
- DIN Standard 18271
- Gray-blue color
Specifications
30mm | |
Blue | |
32 mm | |
Stopper |
50/Pk. | |
26 mm | |
Natural Rubber | |
31D |