Learn More
Fisherbrand™ Natural Rubber Two Hole Stoppers
Used for laboratory purposes.
Brand:  Fisherbrand™ RB071/2H
 View more versions of this product
Product Code. 11547642
  • 17.00€ / Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
Description
Description
Used for laboratory purposes.
  • Made up of natural rubber
  • Red in color
  • Stoppers with two holes
Specifications
Specifications
5 mm
46/55 IRHD
1/Ea.
71 mm
Natural Rubber
For laboratory use
49 mm
Red
78 mm
Tapered Stopper (2 holes)
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Natural Rubber Two Hole Stoppers > Bottom Diameter: 71mm; Top Diameter: 78mm