Learn More
Aalborg™ Needle Valve for Brass Flowmeter
These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes. Aalborg™ Needle Valve for Brass Flowmeter is designed for controlling a broad range of flow rates of liquids and gases.
Brand:  Aalborg™ VCLBB
 View more versions of this product
Product Code. 11862380
  • 79.00€ / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in three conveniently overlapping orifice-needle sizes.
  • These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes.
  • Valves are offered in straight (T) and 90 degree (L) flow patterns. All valves are supplied with 1/8in. FNPT inlet and outlet ports.
  • Valve cartridges are also interchangeable with built-in valves of Aalborg’s series of P, T, S, and G flow meter product line.
  • Bubble tight shutoff
Specifications
Specifications
5000 mL/min.
350mL/min.
Standard Valve
82°C
150 mm Gas/Liquid Flowmeter