Learn More
Cytiva Net Ring
Brand:  Cytiva 28-9663-80
 View more versions of this product
Product Code. 11663224
  • 81.00€ / Each
In Stock
Specifications
Net Ring
5 Pack
For 26 mm HiScale Columns I.D.
Product Title
Cytiva Net Ring > For Use With 26mm HiScale Columns I.D.