missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems NHP XL IL-12 p70, Magnetic Luminex™ Performance Assay

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

Brand:  R&D Systems LNHPXL219

Product Code. 18714324

  • 227.00€ / 1 set
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1 Kit
IL-12 p70/IL-12A
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Primate
3592
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers