missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide adenine dinucleotide phosphate, disodium salt, 98-100%, MP Biomedicals™
Click to view available options
Quantity:
50 mg
100 mg
250 mg
500 mg
1 g
5 g
Specifications
Specifications
Color | White |
Assay Percent Range | 98-100% |
Quantity | 1 g |
Formula Weight | 787.4 |
Physical Form | Powder |
Chemical Name or Material | β-Nicotinamide adenine dinucleotide phosphate, disodium salt and monopotassium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction