Learn More
Nirvanol-d5, Honeywell Fluka™ Product Code.: 15910252
Nirvanol-d5, Honeywell Fluka™
Product Code. 15910252 Shop All Honeywell Fluka Products
10 mg
Click to view available options
Quantity:
10 mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15910252
Brand: Honeywell Fluka R151510MG
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
CAS 119458-27-4
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) Hydantoins
Physical Form Solid
Grade Analytical Standard
Product Content Correction
Product Title
Nirvanol-d5, Honeywell Fluka™ >