Learn More
NISCH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93017-0.02ml
Description
NISCH Polyclonal antibody specifically detects NISCH in Human, Mouse samples. It is validated for Western Blot
Specifications
NISCH | |
Polyclonal | |
Western Blot 1:200 - 1:2000 | |
hIRAS, I-1 receptor candidate protein, I-1FLJ14425, I1R, I1R candidate protein, Imidazoline receptor 1, imidazoline receptor antisera selected, Imidazoline receptor antisera-selected protein, imidazoline receptor candidate, Imidazoline-1 receptor, Imidazoline-1 receptor candidate protein, IR1, IRASFLJ40413, KIAA0975FLJ90519, nischarin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human NISCH (NP_009115.2). SLPQPILSNQGIMFVQEEALASSLSSTDSLTPEHQPIAQGCSDSLESIPAGQAASDDLRDVPGAVGGASPEHAEPEVQVVPGSGQIIFLPFTCIGYTATNQDFIQRLSTLIRQAIERQLPAWIEAANQREEGQGEQGEEEDEEEEEEEDVAENRYFEMGPPDVEEEEGGGQGEEEEEEEEDEEAEEERLALEWALGADEDF | |
0.02 mL | |
Signal Transduction | |
11188 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS (pH 7.3), 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.