Learn More
Ansell™ BioClean™ Clearview™ Autoclavable Goggles Product Code.: 15208108
Add to basket
Ansell™ BioClean™ Clearview™ Autoclavable Goggles
BIOPHARMA
Product Code. 15208108
Goggles
Click to view available options
Goggles
Product Code. 15208108
Brand: Ansell™ BCAG
Add to basket
Add to basket
Constructed in super-soft thermoplastic rubber to provide unrivalled comfort, BioClean Clearview™ Autoclavable Cleanroom Goggles feature upper vents, indirect lower vents, and have a toughened anti-scratch polycarbonate lens. Provided non-sterile. To be autoclaved at 121°C/250°F for 30 minutes
  • Non-Sterile and Sealed in a PE Bag to maintain cleanliness
  • Upper & Indirect Ventilation holes
  • EN 166:2001 Optical Class 1
  • Toughened Anti-Scratch Polycarbonate lens
  • Tested to over 40 Autoclave Cycles
  • Super-soft thermoplastic provides unrivaled comfort
  • Processed and packed in our NEBB Certified ISO Class 4 (Class 10) Cleanroom
  • Replacement lens available (BCAG-SL 19-668-191)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001:2015, (EU)2016/425, EN 166:2001
Lens Coating Scratch-resistant
Product Type Autoclavable Goggles
No. per Case 12/Cs.
No. per Pack 1 Ea.
Ventilation Details Indirect base ventilation
Material Polycarbonate
  • Nitritex Bioclean autoclavable goggles
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Nitritex Bioclean autoclavable goggles
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number24-2995 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog NumberBCAG
    Lot Number24-1606 Certificate TypeCertificate of Compliance Date18/02/2025 Catalog NumberBCAG
    Lot Number24-0061 Certificate TypeCertificate of Compliance Date18/02/2025 Catalog NumberBCAG
    Lot Number23-1250 Certificate TypeCertificate of Compliance Date18/12/2024 Catalog NumberBCAG
    Lot NumberG32312005 Certificate TypeCertificate of Compliance Date23/01/2024 Catalog NumberBCAG
    • Multiple Manufacturers
      Save up to 60% on gloves, apparel, and more
      Promo Code: 24131
      Expires2025-06-08
      New Offer
      Shop Now
    Product Content Correction
    Product Title
    Ansell™ BioClean™ Clearview™ Autoclavable Goggles >