Learn More
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
Brand:  Aalborg™ FT-022-06-SA-VN
 View more versions of this product
Product Code. 11852430
  • 73.00€ / Each
Estimated Shipment: 04-02-2025
Description
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Specifications
200 mL/min.
Nitrogen Direct Flow Tube
65 mm