missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo B receptor Rabbit anti-Human, Mouse, Rat, Clone: S01-3K8, Novus Biologicals™
Rabbit Monoclonal Antibody
290.00€ - 588.00€
Specifications
Antigen | Nogo B receptor |
---|---|
Clone | S01-3K8 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18308606
|
Bio-Techne
NBP3-15069-25UL |
25 μg |
290.00€
25µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392217
|
Bio-Techne
NBP3-15069-100UL |
100 μg |
588.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
Nogo B receptor Monoclonal antibody specifically detects Nogo B receptor in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Nogo B receptor | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
C6orf68MGC117249, chromosome 6 open reading frame 68, MGC:7199, MGC7199, NgBR, nogo-B receptor, Nuclear undecaprenyl pyrophosphate synthase 1 homolog, nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae) | |
A synthetic peptide of human Nogo B receptor (Uniprot#: Q96E22) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S01-3K8 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
116150 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title