Learn More
Socorex™ Pipette Nozzle Protection Filter
Introduce the Socorex™ Pipette Nozzle Protection Filter to protect the pipette from liquid fill or contamination. Made of polypropylene fiber, the filter offers maximal protection and safety from contamination.
Brand: Socorex™ 322.10
This item is not returnable.
View return policy
Description
- Polypropylene fiber
- For 5mL and 10mL Acura™ manual and Acura™ electro
- Non sterile
- Non autoclavable.
Order Info
ForUseWithEquipment