missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Midi

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Midi Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.

Brand:  Macherey-Nagel™ 740410.2

48.69 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 11975442

  • 52.55€ / 2 reactions
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Features
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design –  Midi prep in 30 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 400μg (Midi)
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications

Specifications

Midi Gravity-flow Column
Anion Exchange Chromatography Technology
2
400µg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <

SDS
Documents

Documents

Certificates
Special Offers

Special Offers