missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoCard™
Brand: Macherey-Nagel™ 740403.100
This item is not returnable.
View return policy
Specifications
18°C to 25°C | |
Impregnated Specialized Filter Paper Technology | |
100 | |
< 200μL |
Filter Card | |
Nucleo Card | |
Nucleo Card |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction