Learn More
Macherey-Nagel Bioanalysis™ NucleoMag 96 Blood
Designed for the rapid parallel purification of genomic DNA from whole blood in a 96-well format.
Brand: Macherey-Nagel Bioanalysis™ 744500.1
Description
After lysis of the blood cells genomic DNA is selectively bound to the NucleoMag Blood beads. Contaminants are washed away and the DNA can be eluted from the beads under low-salt conditions. As the magnetic bead technology provides a closed system (no transfer of sample from one well to another is necessary during the preparation) the risk for cross-contamination is greatly reduced. The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg. NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag B-Beads, elution plate (u-bottom), buffers.
- Magnetic bead technology
- Yield: 1 to 4 μg
- Sample material: 100 μL whole blood
- Elution volume: 25 to 100 μL
- Binding capacity: 0.4 μL/ 1 μL beads
- Preparation time: 120 min/96 preps
- Fragment size: 300 bp to 30 kb
- Format: superparamagnetic beads (size of beads: 1 to 3 μm)
Specifications
Magnetic Bead | |
96 Preps | |
Highly Reactive Superparamagnetic Beads (1 to 3 μm) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.